2,5-二苯基噁唑 99,0 HPLC, N spectrometry for scintillation

供应商: TCI
Warning

Synonyms: PPO

D0902-25G D0902-500G
TCIAD0902-25GEA 0 CNY
TCIAD0902-25G TCIAD0902-500G
2,5-二苯基噁唑 99,0 HPLC, N spectrometry for scintillation
2,5-二苯基噁唑
分子式: C₁₅H₁₁NO
分子量: 221.26 g/mol
沸点: 360
存储条件: Ambient
MDL编号: MFCD00005306
CAS号: 92-71-7
EINECS: 202-181-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b96dcdbb61120b87c52a3c9b5e8bba19e87ac710&vid=af2ab6cb1845b917f9171f581102b6ac401076a5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR