Carbon nanotube multi-walled, Length: 1-2 µm, Ø 20-40 nm

供应商: TCI
Warning

Synonyms: CNT

C2155-1G
TCIAC2155-1GEA 0 CNY
TCIAC2155-1G
Carbon nanotube multi-walled, Length: 1-2 µm, Ø 20-40 nm
Carbon nanotube
分子式: C
分子量: 12.011 g/mol
MDL编号: MFCD00133992
CAS号: 308068-56-6

现在订购

产品技术规格

Appearance Black powder to crystal
Purity (TPO Method) min. 95 %
Diameter 20 to 40 nm
Length 1 to 2 micro_m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR