1,3-二(三甲基甲硅烷基)脲 98,0 N_ASSAY_METHOD

供应商: TCI

Synonyms: BSU, N,N′-二(三甲基甲硅烷基)脲

B1103-25G
TCIAB1103-25GEA 0 CNY
TCIAB1103-25G
1,3-二(三甲基甲硅烷基)脲 98,0 N_ASSAY_METHOD
1,3-二(三甲基甲硅烷基)脲
分子式: C₇H₂₀N₂OSi₂
分子量: 204.42 g/mol
MDL编号: MFCD00008260
CAS号: 18297-63-7
EINECS: 242-177-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bcf77bc9ee732adaad320ea5e4f1ebc83a13a63&vid=4bcf77bc9ee732adaad320ea5e4f1ebc83a13a63&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR