4,4′-双(二甲氨基)硫代二苯甲酮 ≥97.0% (by titrimetric analysis) for spectrophotometry

供应商: TCI
Warning

Synonyms: 4,4'-Bis(dimethylamino)thiobenzophenone, N,N,N',N'-Tetramethyl-4,4'-diaminothiobenzophenone, Thiomichler's ketone

A5201-1G
TCIAA5201-1GEA 0 CNY
TCIAA5201-1G
4,4′-双(二甲氨基)硫代二苯甲酮 ≥97.0% (by titrimetric analysis) for spectrophotometry
4,4′-双(二甲氨基)硫代二苯甲酮

Sensitive spectrophotometric reagent for Au, Ag, Hg and Pd, use for the determination of residual chlorine.

分子式: C₁₇H₂₀N₂S
分子量: 284.43 g/mol
MDL编号: MFCD00040477
CAS号: 1226-46-6
EINECS: 214-962-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e80b7c8aa948490b7a670e6fb6954c145f09ce25&vid=f9474fb2f285aa0c139d13e56cb6468bee487e14&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Red to Dark red to Dark purple powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR