4-Aminophenyl beta-D-Galactopyranoside 98,0 HPLC_ASSAY_METHOD

供应商: TCI

Synonyms: (2S,3R,4S,5R,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A1413-1G
TCIAA1413-1GEA 0 CNY
TCIAA1413-1G
4-Aminophenyl beta-D-Galactopyranoside 98,0 HPLC_ASSAY_METHOD
4-Aminophenyl beta-D-Galactopyranoside
分子式: C₁₂H₁₇NO₆
分子量: 271.27 g/mol
MDL编号: MFCD00067362
CAS号: 5094-33-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D -37.0 to -44.0 deg (C=1, MeOH)
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR