氯化钾 3 mol/l (3 N) aqueous solution, Supelco®

供应商: Merck

Synonyms: Potassiumchloride

MQ100

60135-250ML
SUPL60135-250MLEA 0 CNY
SUPL60135-250ML
氯化钾 3 mol/l (3 N) aqueous solution, Supelco®
氯化钾
分子式: KCl
分子量: 74.55 g/mol
沸点: 1420
溶点: 773
密度: 1.13 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011360
CAS号: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c5fadab77938903748de01081f452837fdff738&vid=294a60d21de1a5994ae6d94d5fc36f450ad8808d&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR