(±)-α-生育酚

供应商: Merck

Synonyms: 维生素 E, DL-α-生育酚

47783
SUPL47783EA 0 CNY
SUPL47783
(±)-α-生育酚
(±)-α-生育酚
分子式: C₂₉H₅₀O₂
分子量: 430.71 g/mol
MDL编号: MFCD00072051
CAS号: 10191-41-0
EINECS: 233-466-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da15a597e3aa562597de5daeca96176dcd44e2a1&vid=da15a597e3aa562597de5daeca96176dcd44e2a1&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR