Silica gel, Davisil® Grade 923 pore size 30 Å, 100-200 mesh, high_purity, Supelco®

供应商: Merck

214477-250G
SUPL214477-250GEA 0 CNY
SUPL214477-250G
Silica gel, Davisil® Grade 923 pore size 30 Å, 100-200 mesh, high_purity, Supelco®
Silica gel
分子式: SiO₂
分子量: 60.08 g/mol
沸点: 2230 °C
溶点: > 1600 °C
存储条件: Ambient
MDL编号: MFCD00011232
CAS号: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff484c390424ecbc604290464e00fc361ea19178&vid=ab04e2801dffc54a5f5c0faae685b6bb0ed3cf74&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR