Organic reference standard, Lutein, analytical standard, Supelco®
供应商: Merck
07168-1MG 07168-5MG
SUPL07168-1MGEA 0 CNY
SUPL07168-1MG SUPL07168-5MG
Organic reference standard, Lutein, analytical standard, Supelco®
标准品 Organic Standards
Lutein is a yellow dihydroxylated carotenoid which is found to be a common constituent in many dietary supplements.
Lutein may be used as a reference standard for the determination of lutein in processed foods and supplements, in tomato juice, in human plasma and in leafy vegetables by high performance liquid chromatography-photodiode array detection (HPLC-PDA) and in fish eggs by HPLC coupled with electrospray ionisation (ESI) and tandem mass spectrometry (MS/MS)
Dietary carotenoid with no vitamin A potency. Increases macular pigment concentration in the eye and may improve visual function.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b61adf97828a0ca91c1b952b34df2eebd287c689&vid=b61adf97828a0ca91c1b952b34df2eebd287c689&prid=web&clid=SAPDAM