Microcentrifuge tubes, 0,6 ml and 1,5 ml, Cliklok™
供应商: SIMPORT SCIENTIFIC
T330-8N T330-7G T330-7B T330-7N T330-7Y T330-6N T330-7AM
SIMPT330-8NEA 0 CNY
SIMPT330-8N SIMPT330-7G SIMPT330-7B SIMPT330-7N SIMPT330-7Y SIMPT330-6N SIMPT330-7AM
Microcentrifuge tubes, 0,6 ml and 1,5 ml, Cliklok™
管子 微型离心管
PP, graduated, with flat cap Cliklok™.
  • Can withstand ultra high centrifugation to 20000×g
  • Flat caps can be easily labelled
  • Certified RNase, DNase, pyrogen and DNA-free
  • Cliklok™ sealing caps (lock in two stages) reduce popping where freezing, boiling, storing or shipping is required
Packaging: Microtubes packed in resealable bags.