PCR plates
供应商: SIMPORT SCIENTIFIC
T323-24N T323-24B T323-24G T323-24R T323-24Y T323-48B T323-48G T323-48N T323-48R T323-48Y T323-96B T323-96G T323-96N T323-96R T323-96Y
SIMPT323-24NEA 0 CNY
SIMPT323-24N SIMPT323-24B SIMPT323-24G SIMPT323-24R SIMPT323-24Y SIMPT323-48B SIMPT323-48G SIMPT323-48N SIMPT323-48R SIMPT323-48Y SIMPT323-96B SIMPT323-96G SIMPT323-96N SIMPT323-96R SIMPT323-96Y
PCR plates
微孔板 PCR板
PP. Thin walled PCR plates compatible with 0,2 ml thermal cyclers. The insides of the tubes are smooth and have an inert surface on which enzymes and nucleic acids do not bind.
  • Alphanumeric grid for better identification
  • Flexible plate for better fitting of tubes in the thermal block
  • Larger plates are easily cut into sections of 24, 32 or 48 tubes, but, for more convenience, pre-cut plates are also available with formats of 48 tubes (6×8) and 24 tubes (3×8)
  • The bottom right corner of the plate is cut away to facilitate orientation
  • Certified free of RNase,
More Product Information