Deep well plates, 96-well, Bioblock™
供应商: SIMPORT SCIENTIFIC
T110-6
SIMPT110-6EA 0 CNY
SIMPT110-6
Deep well plates, 96-well, Bioblock™
微孔板 深孔微孔板
PS. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) can withstand centrifugation up to 3000×g. It is not DMSO resistant.
Delivery information: Packed in bags of 4 plates.