Slide boxes, SlideFile™
供应商: SIMPORT SCIENTIFIC
M700-100B M700-100G M700-100P M700-100W M700-100Y
SIMPM700-100BEA 0 CNY
SIMPM700-100B SIMPM700-100G SIMPM700-100P SIMPM700-100W SIMPM700-100Y
Slide boxes, SlideFile™
显微镜幻灯片盒
This impact resistant storage system can hold up to 400 slides per unit, for space efficient storage. All slides are stored upright for easier manipulation.
  • Removable tray inside the storage box for 100 individual numbered slots
  • Transparent hinged cover makes it easy to see the contents of the box at a glance
  • Unique feature with this system to read barcodes without having to remove the slides from the box
W×D×H: 82×245×86 mm
订购须知 Each system includes a slide box and a removable tray.
Delivery information: Index cards numbered from one to 100 forMore Product Information