Tissue processing/embedding cassettes, Unisette™, M505
供应商: SIMPORT SCIENTIFIC
M505-2 M505-3 M505-4 M505-5 M505-6 M505-7 M505-8 M505-9 M505-10 M505-11 M505-12
SIMPM505-2EA 0 CNY
SIMPM505-2 SIMPM505-3 SIMPM505-4 SIMPM505-5 SIMPM505-6 SIMPM505-7 SIMPM505-8 SIMPM505-9 SIMPM505-10 SIMPM505-11 SIMPM505-12
Tissue processing/embedding cassettes, Unisette™, M505
包埋盒 组织处理盒
Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.
  • Flow-through slots maximise fluid exchange and ensure effective drainage
  • Resistant to histological solvents
  • Integral lid
  • Front writing face inclined 35°
  • Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes.