Tween® 20 (Polysorbate), Sigma-Aldrich®

供应商: Merck

Synonyms: Polyoxyethylene sorbitan monolaurate

MQ200

P9416-100ML P9416-50ML
SIALP9416-100MLEA 0 CNY
SIALP9416-100ML SIALP9416-50ML
Tween® 20 (Polysorbate), Sigma-Aldrich®
Tween® 20 (Polysorbate)
分子式: C₅₈H₁₁₄O₂₆
分子量: 1227.53 g/mol
密度: 1.1 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00165986
CAS号: 9005-64-5
EINECS: 500-018-3

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR