乙酸钾, Sigma-Aldrich®

供应商: Merck

Synonyms: Acetic acid potassium salt

M-Clarity™ quality level = MQ200

P1222-500G
SIALP1222-500GEA 0 CNY
SIALP1222-500G
乙酸钾, Sigma-Aldrich®
乙酸钾
分子式: CH₃COOK
分子量: 98.14 g/mol
溶点: 304
密度: 1.57 g/cm³ (25 °C)
闪点: >250 °C
存储条件: Ambient
MDL编号: MFCD00012458
CAS号: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR