α-酮戊二酸, Sigma-Aldrich®

供应商: Merck
Danger

Synonyms: 2-氧化戊二酸, 2-氧代戊二酸

MQ200

K1128-25G
SIALK1128-25GEA 0 CNY
SIALK1128-25G
α-酮戊二酸, Sigma-Aldrich®
α-酮戊二酸
分子式: HOOCCH₂CH₂COCOOH
分子量: 146.1 g/mol
沸点: 323
溶点: 112…116 °C
存储条件: Refrigerator
MDL编号: MFCD00004165
CAS号: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR