肌醇, Sigma-Aldrich®

供应商: Merck

Synonyms: 环己六醇, 肌糖, meso-肌醇

M-Clarity™ quality level = MQ300

I7508-50G
SIALI7508-50GEA 0 CNY
SIALI7508-50G
肌醇, Sigma-Aldrich®
肌醇
分子式: C₆H₁₂O₆
分子量: 180.16 g/mol
沸点: 291
溶点: 210
密度: 1.752 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00077932
CAS号: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR