Chicken native albumin (from egg)

供应商: Merck

A5503-1G
SIALA5503-1GEA 0 CNY
SIALA5503-1G
Chicken native albumin (from egg)
蛋白质

现在订购

规格

Protein/peptide name Albumin
Environmentally Preferable
Protein/peptide type Native
Species Chicken
Source Egg
CAS No. 9006-59-1
存储条件 Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR