2′-氨基苯乙酮, Sigma-Aldrich®

供应商: Merck
Warning

Synonyms: 2-乙酰苯胺

M-Clarity™ quality level = MQ100

A37804-5G
SIALA37804-5GEA 0 CNY
SIALA37804-5G
2′-氨基苯乙酮, Sigma-Aldrich®
2′-氨基苯乙酮
分子式: C₈H₉NO
分子量: 135.17 g/mol
沸点: 252
溶点: 20
密度: 1.12 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00007717
CAS号: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR