碘代乙酰胺, Sigma-Aldrich®

供应商: Merck
Warning

Synonyms: 2-Iodoacetamide

MQ200

A3221-10VL
SIALA3221-10VLEA 0 CNY
SIALA3221-10VL
碘代乙酰胺, Sigma-Aldrich®
碘代乙酰胺
分子式: ICH₂CONH₂
分子量: 184.96 g/mol
沸点: 266
溶点: 91…94 °C
存储条件: Refrigerator
MDL编号: MFCD00008028
CAS号: 144-48-9
EINECS: 205-630-1
UN号: 2811
ADR: 6.1 III

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR