正戊烷磺酸钠, Sigma-Aldrich®

供应商: Merck

Synonyms: 1-戊烷磺酸

76955-50G
SIAL76955-50GEA 0 CNY
SIAL76955-50G
正戊烷磺酸钠, Sigma-Aldrich®
正戊烷磺酸钠
分子式: C₅H₁₁NaO₃S·1H₂O
分子量: 192.21 g/mol
存储条件: Ambient
MDL编号: MFCD00007541
CAS号: 207605-40-1
EINECS: 245-208-4

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR