乙醛, anhydrous ≥99.5% (by GC), puriss. p.a., Sigma-Aldrich®

供应商: Merck
Danger

Synonyms: 醋醛

M-Clarity™ quality level = MQ100

00070-500ML 00070-100ML
SIAL00070-500MLEA 0 CNY
SIAL00070-500ML SIAL00070-100ML
乙醛, anhydrous ≥99.5% (by GC), puriss. p.a., Sigma-Aldrich®
乙醛
分子式: CH₃CHO
分子量: 44.05 g/mol
沸点: 21 °C (lit.)
溶点: –125 °C (lit.)
密度: 0.785 g/mL (25 °C) (lit.)
闪点: –27 °C
存储条件: Refrigerator
MDL编号: MFCD00006991
CAS号: 75-07-0
EINECS: 200-836-8
UN号: 1089
ADR: 3 I
Merck Index: 12,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR