Anti-F4/80 Rat Monoclonal Antibody [clone: Cl:A3-1]

供应商: SEROTEC
MCA497G
SERTMCA497GEA 0 CNY
SERTMCA497G
Anti-F4/80 Rat Monoclonal Antibody [clone: Cl:A3-1]
抗体
Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: Cl:A3-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR