Disinfectant wipes for surfaces, perform® classic wipes EP
供应商: SCHUELKE MAYR
70000186
SCUE70000186EA 0 CNY
SCUE70000186
Disinfectant wipes for surfaces, perform® classic wipes EP
擦拭巾 一次性湿巾和毛巾 Disinfectant Wipes
Ready to use impregnated wipes for disinfecting surfaces.
  • Fast, effective
  • Colour- and perfume-free
  • Tested according to European norms (EN)
For disinfection of surfaces in areas where fast microbiological effectiveness is required.
100 g active agent solution contains:
25 g Ethanol (94 %), 35 g Propan-1-ol.
Packaging: One box contains: 200 ready to use impregnated wipes.
Caution: Use biocides safely. Always read the label and product information before use. Do not use on surfaces that are particularly sensitive to alcohol, such as acrylic glass.