Bottles, clear or amber glass, DURAN® PURE
供应商: DWK LIFE SCIENCES
818062402 818063604 818061406 818011401 818012406 818013608 818014407 818015403 818016305 818017301 818018606 818019105 818064403 818065408 818066301 818067306 818068602 818069101
SCOT818062402EA 0 CNY
SCOT818062402 SCOT818063604 SCOT818061406 SCOT818011401 SCOT818012406 SCOT818013608 SCOT818014407 SCOT818015403 SCOT818016305 SCOT818017301 SCOT818018606 SCOT818019105 SCOT818064403 SCOT818065408 SCOT818066301 SCOT818067306 SCOT818068602 SCOT818069101
Bottles, clear or amber glass, DURAN® PURE
玻璃瓶
DURAN® PURE bottles clear or amber, with GL thread and dust cover. DURAN® PURE is a range of primary packaging products dedicated to the specific requirements and needs of the pharmaceutical industry.
  • Made from DURAN® borosilicate glass 3.3 Type I
  • Protective dust cover is applied in-line at the end of the manufacturing process while the glass still has a very high level of cleanness
  • Lot specific certificate of conformance is available via 'Retrace Code' internet download (includes USP, Ph. Eur., JP analysis per lot)
  • 100% lot traceability of DURAN® PURE
More Product Information