Stopcock, single way, DURAN®
供应商: DWK LIFE SCIENCES
286442003 286442809 286443008 286443805 286444201 286444407 286446005 286446605 286446802 286447001 286445403
SCOT286442003EA 0 CNY
SCOT286442003 SCOT286442809 SCOT286443008 SCOT286443805 SCOT286444201 SCOT286444407 SCOT286446005 SCOT286446605 SCOT286446802 SCOT286447001 SCOT286445403
Stopcock, single way, DURAN®
旋塞阀
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
Straight bore.