Clamps for conical standard ground joints, KECK™, DURAN®
供应商: DWK LIFE SCIENCES
286336109 286336203 286336306 286336409 286336503 286336606 286336709 286336803 286336906
SCOT286336109EA 0 CNY
SCOT286336109 SCOT286336203 SCOT286336306 SCOT286336409 SCOT286336503 SCOT286336606 SCOT286336709 SCOT286336803 SCOT286336906
Clamps for conical standard ground joints, KECK™, DURAN®
夹具 NS联合卡箍
POM. These clips made of temperature resistant polyacetal (POM) are easy, safe and can be fitted quickly.
  • Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape