Tubing connectors, complete with screw cap, straight, DURAN®
供应商: DWK LIFE SCIENCES
286221806
SCOT286221806EA 0 CNY
SCOT286221806
Tubing connectors, complete with screw cap, straight, DURAN®
接头
Screw cap PTB, red, with white PP straight hose connection.
  • Very good chemical resistance
  • Very good temperature resistance
With silicone seal (VMQ).