Stirrer bearing with screw thread DURAN® KPG®
供应商: DWK LIFE SCIENCES
247500803 247500906
SCOT247500803EA 0 CNY
SCOT247500803 SCOT247500906
Stirrer bearing with screw thread DURAN® KPG®
搅拌器 Stirring Tools
DURAN®, clear borosilicate glass 3.3.
  • Interchangeable
  • With standard ground joint and GL screw thread tube
  • Ground and polished surface