Reaction vessels, cylindrical, with flat flange, DURAN®
供应商: DWK LIFE SCIENCES
243902408 243903601 243904409 243905405 243906307 243907106 243907603 243908608
SCOT243902408EA 0 CNY
SCOT243902408 SCOT243903601 SCOT243904409 SCOT243905405 SCOT243906307 SCOT243907106 SCOT243907603 SCOT243908608
Reaction vessels, cylindrical, with flat flange, DURAN®
反应容器 圆柱形反应容器
DURAN®, borosilicate glass 3.3, clear, with round bottom.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat flange (DN) with groove for vacuum seal. Maximum operating pressure at +250 °C: 2,5 bar for 100 and 250 ml; 1,5 bar for 500, 1000 and 2000 ml; 1,0 bar for 4000 and 6000 ml; and 0,5 bar for 10000 ml.
*Vessel cylindrical throughout and has a flat bottom.
Caution: Important: At a maximum operating temperature of +250 °C and at permissibleMore Product Information