Bulb pipettes, class B, DURAN®
供应商: DWK LIFE SCIENCES
243370108 243370202 243370708 243370802 243371207 243371404 243371704 243372409
SCOT243370108EA 0 CNY
SCOT243370108 SCOT243370202 SCOT243370708 SCOT243370802 SCOT243371207 SCOT243371404 SCOT243371704 SCOT243372409
Bulb pipettes, class B, DURAN®
移液管和移液器 球管
Soda-lime glass (AR-Glas®).
  • Marks and inscription in brown
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 648.
Colour code complies to DIN 12621.
*no bulb