Bulb condenser, Allihn type, DURAN®
供应商: DWK LIFE SCIENCES
242527104 242527207 242528109 242528203
SCOT242527104EA 0 CNY
SCOT242527104 SCOT242527207 SCOT242528109 SCOT242528203
Bulb condenser, Allihn type, DURAN®
冷凝器
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With core and jacket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
The bulb condenser has a greater cooling surface than the Liebig condenser and thus higher cooling capacity.