Distillation columns, Vigreux, with glass jacket, DURAN®
供应商: DWK LIFE SCIENCES
242407104 242407207 242408709 242408803
SCOT242407104EA 0 CNY
SCOT242407104 SCOT242407207 SCOT242408709 SCOT242408803
Distillation columns, Vigreux, with glass jacket, DURAN®
蒸馏组件 蒸馏柱
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground joints, complete with slide-on glassjacket.