DURAN® Flasks, Round Bottom, with Two Necks and Standard Gound Joints
供应商: DWK LIFE SCIENCES
241833609 241833703 241835601
SCOT241833609EA 0 CNY
SCOT241833609 SCOT241833703 SCOT241835601
DURAN® Flasks, Round Bottom, with Two Necks and Standard Gound Joints
烧瓶 圆底烧瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one parallel side neck. With matt white labelling area.
*non-DIN ISO size.
Certifications: DIN 12394.