Dropping funnels, cylindrical, without pressure balance, DURAN®
供应商: DWK LIFE SCIENCES
241242008 241242402 241242505 241243604 241243707 241244403 241244609 241245605
SCOT241242008EA 0 CNY
SCOT241242008 SCOT241242402 SCOT241242505 SCOT241243604 SCOT241243707 SCOT241244403 SCOT241244609 SCOT241245605
Dropping funnels, cylindrical, without pressure balance, DURAN®
漏斗 滴液漏斗
DURAN®, borosilicate glass 3.3, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible divisions for approximate volume reading.
DIN ISO 4800.
* non-DIN ISO size