Laboratory bottles, narrow neck, with thread, DURAN®
供应商: DWK LIFE SCIENCES
218011404 218011704 218012409 218013602 218014401 218015406 218016308 218017304 218018609 218019108 218018806 218012906 218015106 218016908 218010802
SCOT218011404EA 0 CNY
SCOT218011404 SCOT218011704 SCOT218012409 SCOT218013602 SCOT218014401 SCOT218015406 SCOT218016308 SCOT218017304 SCOT218018609 SCOT218019108 SCOT218018806 SCOT218012906 SCOT218015106 SCOT218016908 SCOT218010802
Laboratory bottles, narrow neck, with thread, DURAN®
玻璃瓶
DURAN®, borosilicate glass 3.3, clear, without screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduation and DIN thread. Without pouring ring or screw cap.
With printed retrace code (batch identification).
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominalMore Product Information