Sedimentation cones, Imhoff type, DURAN®
供应商: DWK LIFE SCIENCES
214015403
SCOT214015403EA 0 CNY
SCOT214015403
Sedimentation cones, Imhoff type, DURAN®
漏斗 沉淀锥形瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • Very good chemical resistance
  • Without stopcock
The conical sedimentation cone with graduation according to Imhoff allows a quantification of sediments from a suspension (e.g. in waste water).
Certifications: According to DIN 12672.