Reagent/centrifuge tubes, flat base, with screw cap
供应商: SARSTEDT
71.9923.610
SARS71.9923.610EA 0 CNY
SARS71.9923.610
Reagent/centrifuge tubes, flat base, with screw cap
管子 离心管
PP, translucent, PS or PC, transparent with PP screw caps
  • Non sterile or sterile
  • Caps enclosed or assembled available
  • With or without graduation
Universal tubes for a wide range of applications.