Precision balances, verified models with EC-type approval certificate (-0CE), Cubis®
供应商: SARTORIUS BALANCES
MSE6202S-1CE-D
SARBMSE6202S-1CE-DEA 0 CNY
SARBMSE6202S-1CE-D
Precision balances, verified models with EC-type approval certificate (-0CE), Cubis®
天平
The Cubis® range delivers quality weighing together with a high degree of flexibility without having to invest in expensive equipment that has some features that may never be required. The MSE control unit has a black and white basic backlit LCD display and is ideal for straightforward high accuracy weighing.
  • Removable control units and display
  • RS232 and USB interfaces are included
  • Easy to upgrade separate modules
  • Internal, IsoCAL; fully automatic calibration and adjustment