Disposable forceps, SteriWare®
供应商: SAMPLING SYSTEMS
8025H-115S 8025H-115
SAMP8025H-115SEA 0 CNY
SAMP8025H-115S SAMP8025H-115
Disposable forceps, SteriWare®
镊子
These diposable tweezers made from 100% virgin HDPE are ideal for picking up items up to 25 mm. Its special dual action design means it can pick up both small and large items.
  • Made from FDA and EC conforming material
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • Gamma irradiated (SAL 10⁻⁶)
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
Packaging: Manufactured and packed in a class 100000 cleanroom environment.More Product Information