Spatulas, Micro, Disposable, SteriWare®
供应商: SAMPLING SYSTEMS
8015A-170 8015A-170S 8015V-170S 8015H-170 8015N-170 8015N-170S 8015NR-170S 8015H-170S 8015V-170
SAMP8015A-170EA 0 CNY
SAMP8015A-170 SAMP8015A-170S SAMP8015V-170S SAMP8015H-170 SAMP8015N-170 SAMP8015N-170S SAMP8015NR-170S SAMP8015H-170S SAMP8015V-170
Spatulas, Micro, Disposable, SteriWare®
样品勺
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
  • Moulded and packed in a cleanroom
  • Suitable for food contact
  • Available sterile (gamma irradiated)
  • Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.