Chemical spoon with spatula end
供应商: RSG
021.120 021.150 021.210 021.250 021.180 021.300
RSGA021.120EA 0 CNY
RSGA021.120 RSGA021.150 RSGA021.210 RSGA021.250 RSGA021.180 RSGA021.300
Chemical spoon with spatula end
勺子
Stainless steel Remanit 4301, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean