Planetary ball mills, PM 100, PM 100 CM, PM 200 and PM 400
供应商: RETSCH
22.782.0029 22.782.0030
RETS22.782.0029EA 0 CNY
RETS22.782.0029 RETS22.782.0030
Planetary ball mills, PM 100, PM 100 CM, PM 200 and PM 400
研磨机
Planetary mills pulverise and mix soft, medium-hard to extremely hard, brittle and fibrous materials quickly without loss. Examples of applications include minerals, ores, chemicals, ceramics, soils, household and industrial waste and sample preparation for spectral analysis. These mills are suitable for long term trials and continuous use and are used for dry and wet grinding in all industry sectors, where a high degree of reproducibility, fineness, purity and speed is required. The excellent performance is achieved by the high centrifugal forces and very high pulverisation energy generatedMore Product Information