FTA® Elute Cards, Whatman™
供应商: QIAGEN
WB120410 WB120401 WB120411 WB120412
QIAGWB120410EA 0 CNY
QIAGWB120410 QIAGWB120401 QIAGWB120411 QIAGWB120412
FTA® Elute Cards, Whatman™
核酸储存系统
FTA® Elute Cards utilise Whatman FTA® technology that simplifies the handling and processing of nucleic acids. DNA is eluted in an easy step, providing DNA in solution ready for amplification.
  • Samples can be collected, shipped, and preserved at room temperature, eliminating the high costs associated with shipping samples on ice and with laboratory freezer storage
  • Sample processing time to isolate DNA is 15 to 30 minutes, avoiding the need for lengthy and multiple step isolation procedures
  • Sample processing requires a simple hot water elution procedure to
More Product Information