FTA® Cards, Whatman™
供应商: QIAGEN
WB120205 WB120210 WB120211 WB120206 WB120055 WB120056 WB120208 WB120355 WB120356 WB120065 WB120305 WB120306 WB120310 WB120311
QIAGWB120205EA 0 CNY
QIAGWB120205 QIAGWB120210 QIAGWB120211 QIAGWB120206 QIAGWB120055 QIAGWB120056 QIAGWB120208 QIAGWB120355 QIAGWB120356 QIAGWB120065 QIAGWB120305 QIAGWB120306 QIAGWB120310 QIAGWB120311
FTA® Cards, Whatman™
核酸储存系统
For the collection, transport, archiving and isolation of nucleic acids at room temperature. FTA® Cards contain chemicals that lyse cells, denature proteins and protect nucleic acids from nucleases, oxidation and UV damage. The cards rapidly inactivate organisms, including blood-borne pathogens, and prevent the growth of bacteria and other microorganisms. FTA® Cards are available in a variety of configurations. Applications include forensics, transgenics, transfusion medicine, plasmid screening, drug discovery and genomics.
  • Nucleic acids are captured, immobilised, and stabilised for
More Product Information