FTA® reagent and accessories, Whatman™
供应商: QIAGEN
WB120204 WB100003 WB100032 WB100035 WB100036 WB100037
QIAGWB120204EA 0 CNY
QIAGWB120204 QIAGWB100003 QIAGWB100032 QIAGWB100035 QIAGWB100036 QIAGWB100037
FTA® reagent and accessories, Whatman™
核酸储存系统
For collection, storage, processing and shipping FTA® cards.
  • FTA® purification reagent: For purification of nucleic acids stored on FTA® cards
  • Ensures superior quality DNA for PCR or SNP analysis
  • Removes haem, PCR inhibitors and other potential contaminants
  • Non toxic, hypoallergenic, aqueous solution
Sterile foam tipped applicator: For the collection of saliva and buccal cells. Non abrasive foam head is the same size as sample area on indicating FTA® Cards to facilitate sample application.
Sterile omni swab: Non invasive device for collectingMore Product Information