Ultra-Light Bump Cap
供应商: PORTWEST CLOTHING
PS89ORR
PRTWPS89ORREA 0 CNY
PRTWPS89ORR
Ultra-Light Bump Cap
头盔 Bump Caps
Industrial Safety Bump Cap with an ultra-light plastic shell provides a lightweight and comfortable solution against minor head injuries such as bumps and scrapes.
  • Lightweight and comfortable
  • Four-point plastic suspension harness for wearer comfort, safety, and easy cleaning
  • Soft foam sweat band included
  • Buckle size adjustment for easy fitting
  • Retail bag aids presentation for retail sales
Bump cap includes four-point plastic harness with a sweatband and adjustable buckle size (500 to 600 mm). Ventilation holes in the shell for extra airflow. Easy toMore Product Information