二甲基亚砜 99,5 LC-MS, Pierce™

供应商: Thermo Fisher Scientific

Synonyms: 甲基亚砜, DMSO

85190
PIER85190EA 0 CNY
PIER85190
二甲基亚砜 99,5 LC-MS, Pierce™
二甲基亚砜

The Pierce™ Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.


  • High-purity solvent for various protein, peptide, and amino acid applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps
  • Suitable for HPLC detection


Caution: For Research Use Only. Not for use in diagnostic procedures.

分子式: (CH₃)₂SO
分子量: 78.14 g/mol
沸点: 189
溶点: 18.5
密度: 1.101 g/cm³ (20 °C)
闪点: 87
MDL编号: MFCD00002089
CAS号: 67-68-5
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

现在订购

产品技术规格

Visual Appearance Clear, Colorless liquid, Free of particulate matter
Purity ≥ 99.5%
Water Content ≤ 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR