Pierce™ ABC Staining Kits
供应商: Thermo Fisher Scientific
32020
PIER32020EA 0 CNY
PIER32020
Pierce™ ABC Staining Kits
蛋白连接和标记系统
ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.
  • Very low background staining
  • Rapid avidin-biotin interaction
Standard Kits containing only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-sensitive ABC Peroxidase Staining Kits enable more dilute primary antibodies to be used while producing equal staining intensity. Ultra-sensitive kits are offered as a standard kit and as complete kits, which contain biotinylated secondary antibodies and appropriate blocking serum for use with mouse or rabbit antibodies.